Learn More
Biolegend TotalSeq™-C0141 anti-human CD195 (CCR5)
Supplier:  Biolegend 359137
Encompass_Preferred
TotalSeq™-C0141 anti-human CD195 (CCR5) [J418F1]; Isotype: Rat IgG2b, κ; Reactivity: Human; Apps: PG; Size: 10 ug
Catalog No. 50-207-7824