Learn More
Biolegend TotalSeq™-C0152 anti-human CD223 (LAG-3)
Supplier:  Biolegend 369335
Encompass_Preferred
TotalSeq™-C0152 anti-human CD223 (LAG-3) [11C3C65]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: PG; Size: 10 ug
Catalog No. NC1930235