Learn More
Biolegend TotalSeq™-C0158 anti-human CD134 (OX40)
Supplier:  Biolegend 350035
Encompass_Preferred
TotalSeq™-C0158 anti-human CD134 (OX40) [Ber-ACT35 (ACT35)]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: PG; Size: 10 ug
Catalog No. NC1930236