Learn More
Biolegend TotalSeq™-C0168 anti-human CD57 Recombinant
Supplier:  Biolegend 393321
Encompass_Preferred
TotalSeq™-C0168 anti-human CD57 Recombinant [QA17A04]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: PG; Size: 10 ug
Catalog No. 50-207-9000