Learn More
Biolegend TotalSeq™-C0255 anti-human Hashtag 5
Supplier:  Biolegend 394669
Encompass_Preferred
TotalSeq™-C0255 anti-human Hashtag 5 [LNH-94; 2M2]; Isotype: Mouse IgG1, κ (all clones); Reactivity: Human; Apps: PG; Size: 10 ug
Catalog No. 50-207-9135