Learn More
Biolegend TotalSeq™-C0591 anti-human LOX-1
Supplier:  Biolegend 358613
Encompass_Preferred
TotalSeq™-C0591 anti-human LOX-1 [15C4]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: PG; Size: 10 ug
Catalog No. 50-207-7769