Learn More
Biolegend TotalSeq™-C0821 anti-human CD164
Supplier:  Biolegend 324813
Encompass_Preferred
TotalSeq™-C0821 anti-human CD164 [67D2]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: PG; Size: 10 ug
Catalog No. 50-207-5688