Learn More
Biolegend TotalSeq™-C0864 anti-human CD352 (NTB-A)
Supplier:  Biolegend 317213
Encompass_Preferred
TotalSeq™-C0864 anti-human CD352 (NTB-A) [NT-7]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus; Apps: PG; Size: 10 ug
Catalog No. NC1927479