Learn More
Biolegend TotalSeq™-C0897 anti-human CD23
Supplier:  Biolegend 338525
Encompass_Preferred
TotalSeq™-C0897 anti-human CD23 [EBVCS-5]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: PG; Size: 10 ug
Catalog No. 50-207-6347