Learn More
Biolegend TotalSeq™-C0902 anti-human CD328 (Siglec-7)
Supplier:  Biolegend 339213
Encompass_Preferred
TotalSeq™-C0902 anti-human CD328 (Siglec-7) [6-434]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: PG; Size: 10 ug
Catalog No. 50-207-6393