Learn More
Biolegend TotalSeq™-C0922 anti-human CD4 [OKT4]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: PG; Size: 10 ug
Supplier:  Biolegend 317455
Encompass_Preferred
TotalSeq™-C0922 anti-human CD4 [OKT4]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: PG; Size: 10 ug
Catalog No. 50-237-4622