Learn More
Biolegend TotalSeq™-C0966 PE Streptavidin
Supplier:  Biolegend 405161
Encompass_Preferred
TotalSeq™-C0966 PE Streptavidin Reactivity: Human, Mouse, Rat, All Species; Apps: PG; Size: 10 ug
Catalog No. 50-213-2016