Learn More
Biolegend TotalSeq™-C1027 anti-human PSMA (FOLH1) [LNI-17]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: PG; Size: 10 ug
Supplier:  Biolegend 342517
Encompass_Preferred
TotalSeq™-C1027 anti-human PSMA (FOLH1) [LNI-17]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: PG; Size: 10 ug
Catalog No. 50-237-6711