Learn More
Biolegend TotalSeq™-C1262 anti-human CD132 (common γ chain) [TUGh4]; Isotype: Rat IgG2b, κ; Reactivity: Human; Apps: PG; Size: 10 ug
Supplier:  Biolegend 338611
Encompass_Preferred
TotalSeq™-C1262 anti-human CD132 (common γ chain) [TUGh4]; Isotype: Rat IgG2b, κ; Reactivity: Human; Apps: PG; Size: 10 ug
Catalog No. 50-238-5158