Learn More
Biolegend TotalSeq™-A0061 anti-human CD117 (c-kit)
Supplier:  Biolegend 313241
Encompass_Preferred
TotalSeq™-A0061 anti-human CD117 (c-kit) [104D2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus, Cattle (Bovine, Cow); Apps: PG; Size: 10 ug
Catalog No. 50-207-3617