Learn More
Biolegend TotalSeq™-A0064 anti-human CD123
Supplier:  Biolegend 306037
Encompass_Preferred
TotalSeq™-A0064 anti-human CD123 [6H6]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Rhesus; Apps: PG; Size: 10 ug
Catalog No. 50-207-3256