Learn More
Biolegend TotalSeq™-A0076 anti-mouse/human CD15 (SSEA-1)
Supplier:  Biolegend 125615
Encompass_Preferred
TotalSeq™-A0076 anti-mouse/human CD15 (SSEA-1) [MC-480]; Isotype: Mouse IgM, κ; Reactivity: Mouse, Cross-Reactivity: Human; Apps: PG; Size: 10 ug
Catalog No. 50-207-0684