Learn More
Biolegend TotalSeq™-A0104 anti-mouse CD102
Supplier:  Biolegend 105613
Encompass_Preferred
TotalSeq™-A0104 anti-mouse CD102 [3C4 (MIC2/4)]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: PG; Size: 10 ug
Catalog No. 50-206-9893