Learn More
Biolegend TotalSeq™-A0245 anti-human CD106
Supplier:  Biolegend 305813
Encompass_Preferred
TotalSeq™-A0245 anti-human CD106 [STA]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: PG; Size: 10 ug
Catalog No. 50-207-3240