Learn More
Biolegend TotalSeq™-A0247 anti-human CD267 (TACI)
Supplier:  Biolegend 311913
Encompass_Preferred
TotalSeq™-A0247 anti-human CD267 (TACI) [1A1]; Isotype: Rat IgG2a, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: PG; Size: 10 ug
Catalog No. 50-207-3539