Learn More
Biolegend TotalSeq™-A0380 anti-rat CD90/mouse CD90.1 (Thy-1.1)
Supplier:  Biolegend 202547
Encompass_Preferred
TotalSeq™-A0380 anti-rat CD90/mouse CD90.1 (Thy-1.1) [OX-7]; Isotype: Mouse IgG1, κ; Reactivity: Rat, Mouse (AKR/J and PL mouse strains). Cross-Reactivity: Rabbit (Lapine), Guinea Pig6; Apps: PG; Size: 10 ug
Catalog No. 50-207-2510