Learn More
Biolegend TotalSeq™-A0402 anti-human CD1a
Supplier:  Biolegend 300133
Encompass_Preferred
TotalSeq™-A0402 anti-human CD1a [HI149]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: PG; Size: 10 ug
Catalog No. 50-207-2649