Learn More
Biolegend TotalSeq™-A0405 anti-human CD284 (TLR4)
Supplier:  Biolegend 312817
Encompass_Preferred
TotalSeq™-A0405 anti-human CD284 (TLR4) [HTA125]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cross-Reactivity: Guinea Pig; Apps: PG; Size: 10 ug
Catalog No. 50-207-3583