Learn More
Biolegend TotalSeq™-A0810 anti-mouse CD138 (Syndecan-1)
Supplier:  Biolegend 142532
Encompass_Preferred
TotalSeq™-A0810 anti-mouse CD138 (Syndecan-1) [281-2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: PG; Size: 10 ug
Catalog No. 50-207-1400