Learn More
Biolegend TotalSeq™-A1009 anti-mouse CD94
Supplier:  Biolegend 105515
Encompass_Preferred
TotalSeq™-A1009 anti-mouse CD94 [18d3]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: PG; Size: 10 ug
Catalog No. 50-206-9891