Learn More
Biolegend TotalSeq™-B0112 anti-mouse CD62L
Supplier:  Biolegend 104465
Encompass_Preferred
TotalSeq™-B0112 anti-mouse CD62L [MEL-14]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: PG; Size: 10 ug
Catalog No. 50-206-9827