Learn More
Biolegend TotalSeq™-B0200 anti-mouse CD86
Supplier:  Biolegend 105057
Encompass_Preferred
TotalSeq™-B0200 anti-mouse CD86 [GL-1]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: PG; Size: 10 ug
Catalog No. 50-206-9870