Learn More
Biolegend TotalSeq™-B0227 anti-mouse CD122 (IL-2Rβ)
Supplier:  Biolegend 105913
Encompass_Preferred
TotalSeq™-B0227 anti-mouse CD122 (IL-2Rβ) [5H4]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: PG; Size: 10 ug
Catalog No. 50-206-9919