Learn More
Biolegend TotalSeq™-B0369 anti-human CD29
Supplier:  Biolegend 303033
Encompass_Preferred
TotalSeq™-B0369 anti-human CD29 [TS2/16]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cattle (Bovine, Cow), Cynomolgus, Dog (Canine), (Horse reactivity), Rhesus; Apps: PG; Size: 10 ug
Catalog No. 50-207-2940