Learn More
Biolegend TotalSeq™-B0448 anti-mouse CD204
Supplier:  Biolegend 154705
Encompass_Preferred
TotalSeq™-B0448 anti-mouse CD204 [1F8C33]; Isotype: Rat IgG2a; Reactivity: Mouse; Apps: PG; Size: 10 ug
Catalog No. 50-207-2161