Learn More
Biolegend TotalSeq™-B0824 anti-mouse P2X7R
Supplier:  Biolegend 148715
Encompass_Preferred
TotalSeq™-B0824 anti-mouse P2X7R [1F11]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: PG; Size: 10 ug
Catalog No. 50-207-1821