Learn More
Biolegend TotalSeq™-C0367 anti-human CD2
Supplier:  Biolegend 309231
Encompass_Preferred
TotalSeq™-C0367 anti-human CD2 [TS1/8]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: PG; Size: 10 ug
Catalog No. 50-207-3399