Learn More
Biolegend TotalSeq™-C0374 anti-human CD98
Supplier:  Biolegend 315607
Encompass_Preferred
TotalSeq™-C0374 anti-human CD98 [MEM-108]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: PG; Size: 10 ug
Catalog No. 50-207-3702