Learn More
Biolegend TotalSeq™-C0386 anti-human CD28
Supplier:  Biolegend 302963
Encompass_Preferred
TotalSeq™-C0386 anti-human CD28 [CD28.2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Baboon, Capuchin Monkey, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus, Sooty Mangabey, Squirrel Monkey; Apps: PG; Size: 10 ug
Catalog No. 50-207-2927