Learn More
Biolegend TotalSeq™-C0437 anti-mouse/human CD207 (Langerin)
Supplier:  Biolegend 144207
Encompass_Preferred
TotalSeq™-C0437 anti-mouse/human CD207 (Langerin) [4C7]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse, Human, Cross-Reactivity: Cynomolgus, Rabbit, Swine (Pig, Porcine); Apps: PG; Size: 10 ug
Catalog No. 50-207-1528