Learn More
Biolegend TotalSeq™-C0895 anti-mouse/human Mac-2 (Galectin-3)
Supplier:  Biolegend 125423
Encompass_Preferred
TotalSeq™-C0895 anti-mouse/human Mac-2 (Galectin-3) [M3/38]; Isotype: Rat IgG2a, κ; Reactivity: Mouse, Human; Apps: PG; Size: 10 ug
Catalog No. 50-207-0678