Learn More
Biolegend TotalSeq™-C0898 anti-human Ig light chain λ
Supplier:  Biolegend 316629
Encompass_Preferred
TotalSeq™-C0898 anti-human Ig light chain λ [MHL-38]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Baboon, Cynomolgus, Rhesus; Apps: PG; Size: 10 ug
Catalog No. 50-207-3754