Learn More
Biolegend TotalSeq™-C0989 anti-mouse CD98 (4F2)
Supplier:  Biolegend 128221
Encompass_Preferred
TotalSeq™-C0989 anti-mouse CD98 (4F2) [RL388]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: PG; Size: 10 ug
Catalog No. 50-207-0856