Learn More
Biolegend TotalSeq™-A0135 anti-human CD324 (E-Cadherin)
Supplier:  Biolegend 324125
Encompass_Preferred
TotalSeq™-A0135 anti-human CD324 (E-Cadherin) [67A4]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: PG; Size: 10 ug
Catalog No. 50-207-5638