Learn More
Biolegend TotalSeq™-B1150 anti-human IL-12/IL-23 p40 [C11.5]; Isotype: Mouse IgG1, κ; Reactivity: Human, IL-12/IL-23 p40 subunit (monomer, homodimer and heterodimer IL-12 p35/p40 or IL-23 p19/p40); Apps: ICPG; Size: 10 ug
Supplier:  Biolegend 501825
Encompass_Preferred
TotalSeq™-B1150 anti-human IL-12/IL-23 p40 [C11.5]; Isotype: Mouse IgG1, κ; Reactivity: Human, IL-12/IL-23 p40 subunit (monomer, homodimer and heterodimer IL-12 p35/p40 or IL-23 p19/p40); Apps: ICPG; Size: 10 ug
Catalog No. 50-237-4699