Learn More
Biolegend TotalSeqTM-A0083 anti-human CD
Catalog No. 502722503
Encompass_Preferred
TotalSeq™-A0083 anti-human CD16 [3G8]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: PG; Size: 10 μg
Catalog No. 50-272-2503 Supplier Biolegend Supplier No. 302061
TotalSeq™-A0083 anti-human CD16 [3G8]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: PG; Size: 10 μg