Learn More
Biolegend TotalSeq™-B0466 anti-Nuclear Pore Complex Proteins Hashtag 16 [MAb414]; Isotype: Mouse IgG1; Reactivity: Vertebrate, Xenopus, Yeast; Apps: PG; Size: 10 ug
Supplier:  Biolegend 682287
Encompass_Preferred
TotalSeq™-B0466 anti-Nuclear Pore Complex Proteins Hashtag 16 [MAb414]; Isotype: Mouse IgG1; Reactivity: Vertebrate, Xenopus, Yeast; Apps: PG; Size: 10 ug
Catalog No. 50-238-8208