Learn More
Biolegend TotalSeqTM-B0909 anti-mouse CD
Catalog No. 502722633
Encompass_Preferred
TotalSeq™-B0909 anti-mouse CD182 (CXCR2) [SA044G4]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: PG; Size: 10 μg
Catalog No. 50-272-2633 Supplier Biolegend Supplier No. 149321
TotalSeq™-B0909 anti-mouse CD182 (CXCR2) [SA044G4]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: PG; Size: 10 μg