Learn More
Biolegend TotalSeqTM-B1461 anti-human CD
Supplier:  Biolegend 360309
Encompass_Preferred
TotalSeq™-B1461 anti-human CD213a2 (IL13Rα2) [47]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: PG; Size: 10 μg
Catalog No. 50-274-0668