Learn More
Biolegend TotalSeq™-B1469 anti-human erbB3/HER-3 [1B4C3]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: PG; Size: 10 μg
Catalog No. 502723837
Encompass_Preferred
TotalSeq™-B1469 anti-human erbB3/HER-3 [1B4C3]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: PG; Size: 10 μg
Catalog No. 50-272-3837 Supplier Biolegend Supplier No. 324715
TotalSeq™-B1469 anti-human erbB3/HER-3 [1B4C3]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: PG; Size: 10 μg