Learn More
Biolegend TotalSeqTM-C0949 anti-mouse CD
Catalog No. 502722690
Encompass_Preferred
TotalSeq™-C0949 anti-mouse CD226 (DNAM-1) [TX42.1]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: PG; Size: 10 μg
Catalog No. 50-272-2690 Supplier Biolegend Supplier No. 133631
TotalSeq™-C0949 anti-mouse CD226 (DNAM-1) [TX42.1]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: PG; Size: 10 μg