Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azure Biosystems TotalStain Q, Nitrocellulose Membranes
SDP

Catalog No. AC2227AZ Shop All Azure Biosystems Products
Change view
Click to view available options
Concentration:
1X
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
AC2227AZ 1X
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC2227AZ Supplier Azure Biosystems Supplier No. AC2227
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorescent total protein stain for nitrocellulose membranes. Sufficient amount for 15 mini-blots. 600 mL kit.

  • Fluorescent Total Protein Stain for Western blots
  • Accurate and quantitative fluorescent total protein stain to perform normalization of Western blots. Blots can be imaged on both camera-based and scanner instruments with green channel or Safe-Dyes channel.
  • TotalStain Q only adds a short 24-minute step to your Western blot protocol, between protein transfer and blocking buffer incubation. It is compatible with subsequent Western blot detection using chemiluminescence or Near Infrared (NIR) fluorescence.
  • Kit contains enough material for 15 membranes

Specifications

Color Clear
Concentration 1X
Detection Method Fluorescense
Format Bottle
Product Type Protein Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.