Learn More
Lab Storage Systems Inc ToteTray Cassette Box 10/cs
SDP
Catalog No. NC1648037
Encompass
Organize and transport your cassettes around the lab. Designed for use in the lab during processing. Set the ToteTray next to the embedding center & organize your cassettes in rows. Handy dividers included for even more organization. Move easily to the microtome.
Catalog No. NC1648037 Supplier Lab Storage Systems Inc Supplier No. BTC30
Organize and transport your cassettes around the lab. Designed for use in the lab during processing. Set the ToteTray next to the embedding center & organize your cassettes in rows. Handy dividers included for even more organization. Move easily to the microtome.