Learn More
Biolegend TotalSeq™-A0219 anti-human CD119 (IFN-γ R α chain)
Supplier:  Biolegend 308607
Encompass_Preferred
TotalSeq™-A0219 anti-human CD119 (IFN-γ R α chain) [GIR-208]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: PG; Size: 10 ug
Catalog No. 50-207-3380