Learn More
Biolegend TotalSeq™-A0400 anti-human CD144 (VE-Cadherin)
Supplier:  Biolegend 348517
Encompass_Preferred
TotalSeq™-A0400 anti-human CD144 (VE-Cadherin) [BV9]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: PG; Size: 10 ug
Catalog No. 50-207-6885